Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127.3 | NP_001118.3 | 16 - 17 | 2354 | 69 |
NM_145730.2 | NP_663782.2 | 15 - 16 | 2333 | 69 | |
GenBank mRNA | AK294406.1 | - | 7 - 8 | 918 | 69 |
AK296567.1 | - | 4 - 5 | 677 | 69 | |
AK299613.1 | - | 7 - 8 | 919 | 69 | |
CT841508.1 | - | 14 - 15 | 2176 | 69 | |
L13939.1 | - | 15 - 16 | 2213 | 69 |