Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127.3 | NP_001118.3 | 5 - 6 | 718 | 76 |
NM_001166019.1 | NP_001159491.1 | 5 - 6 | 718 | 76 | |
NM_145730.2 | NP_663782.2 | 5 - 6 | 718 | 76 | |
GenBank mRNA | BC046242.1 | - | 5 - 6 | 718 | 76 |
BM909530.1 | - | 5 - 6 | 715 | 76 | |
CT841508.1 | - | 4 - 5 | 561 | 76 | |
L13939.1 | - | 4 - 5 | 577 | 76 |