Assay Design
Gene Transcripts
- Gene Symbol
- FAM47E
- Entrez Gene ID
-
100129583
- Gene Name
- family with sequence similarity 47 member E
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 76214040 - 76283783 on Build GRCh38
- Gene Symbol
- FAM47E-STBD1
- Entrez Gene ID
-
100631383
- Gene Name
- FAM47E-STBD1 readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 76251700 - 76311130 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090914.1 |
- |
2 - 3 |
177 |
114 |
AK296418.1 |
- |
3 - 4 |
373 |
114 |
View Full Product Details