Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080510.4 | NP_001073979.3 | 4 - 5 | 106 | |
NM_001206983.2 | NP_001193912.1 | 4 - 5 | 106 | ||
NM_001206984.2 | NP_001193913.1 | 4 - 5 | 106 | ||
NM_001206985.2 | NP_001193914.1 | 3 - 4 | 106 | ||
NM_001206986.2 | NP_001193915.1 | 3 - 4 | 106 | ||
NM_001206987.2 | NP_001193916.1 | 3 - 4 | 106 | ||
NM_001302703.1 | NP_001289632.1 | 5 - 6 | 106 | ||
NM_001302704.1 | NP_001289633.1 | 4 - 5 | 106 | ||
NM_001302705.1 | NP_001289634.1 | 4 - 5 | 106 | ||
NR_038193.2 | 2 - 3 | 106 | |||
XM_006721674.3 | XP_006721737.1 | 4 - 5 | 106 | ||
XM_006721675.1 | XP_006721738.1 | 4 - 5 | 106 | ||
XM_006721676.3 | XP_006721739.1 | 4 - 5 | 106 | ||
XM_006721678.3 | XP_006721741.1 | 4 - 5 | 106 | ||
XM_006721679.3 | XP_006721742.1 | 3 - 4 | 106 | ||
XM_006721680.2 | XP_006721743.1 | 3 - 4 | 106 | ||
XM_011524282.1 | XP_011522584.1 | 4 - 5 | 106 | ||
XM_017024146.1 | XP_016879635.1 | 4 - 5 | 106 | ||
GenBank mRNA | BC033998.2 | - | 3 - 4 | 106 | |
BC106939.2 | - | 2 - 3 | 106 | ||
BU176249.1 | - | 4 - 5 | 106 | ||
BX356072.2 | - | 4 - 5 | 106 | ||
BX403283.2 | - | 4 - 5 | 106 | ||
CN372746.1 | - | 2 - 3 | 106 | ||
HY285382.1 | - | 1 - 2 | 394 | 106 | |
JQ313903.1 | - | 3 - 4 | 106 |