Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000388.3 | NP_000379.2 | 4 - 5 | 1748 | 65 |
NM_001178065.1 | NP_001171536.1 | 4 - 5 | 1814 | 65 | |
XM_005247837.2 | XP_005247894.1 | 2 - 3 | 978 | 65 | |
XM_006713789.3 | XP_006713852.1 | 4 - 5 | 2329 | 65 | |
XM_017007324.1 | XP_016862813.1 | 5 - 6 | 3694 | 65 | |
XM_017007325.1 | XP_016862814.1 | 4 - 5 | 1720 | 65 | |
GenBank mRNA | BC104999.1 | - | 3 - 4 | 1400 | 65 |
BC112236.1 | - | 3 - 4 | 1400 | 65 | |
D50855.1 | - | 3 - 4 | 1404 | 65 | |
S83176.1 | - | 3 - 4 | 1376 | 65 | |
U20759.1 | - | 4 - 5 | 1748 | 65 | |
U20760.1 | - | 4 - 5 | 1814 | 65 |