Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267041.2 | NP_001253970.1 | 13 - 14 | 1514 | 79 |
NM_001267042.2 | NP_001253971.1 | 13 - 14 | 1406 | 79 | |
NM_001282342.1 | NP_001269271.1 | 12 - 13 | 1437 | 79 | |
NM_015396.5 | NP_056211.2 | 15 - 16 | 1683 | 79 | |
XM_006713564.3 | XP_006713627.1 | 14 - 15 | 1602 | 79 | |
XM_006713565.2 | XP_006713628.1 | 15 - 16 | 1643 | 79 | |
XM_006713566.3 | XP_006713629.1 | 13 - 14 | 1497 | 79 | |
XM_006713567.3 | XP_006713630.1 | 14 - 15 | 1602 | 79 | |
XM_011512632.1 | XP_011510934.1 | 14 - 15 | 1550 | 79 | |
XM_011512633.1 | XP_011510935.1 | 14 - 15 | 1604 | 79 | |
XM_017006095.1 | XP_016861584.1 | 14 - 15 | 1404 | 79 | |
XM_017006096.1 | XP_016861585.1 | 13 - 14 | 1497 | 79 | |
XM_017006097.1 | XP_016861586.1 | 13 - 14 | 1350 | 79 | |
XM_017006098.1 | XP_016861587.1 | 12 - 13 | 1296 | 79 | |
XM_017006099.1 | XP_016861588.1 | 13 - 14 | 1387 | 79 | |
XM_017006100.1 | XP_016861589.1 | 14 - 15 | 1602 | 79 | |
GenBank mRNA | AK001721.1 | - | 13 - 14 | 1245 | 79 |
AK001882.1 | - | 6 - 7 | 818 | 79 | |
AK296757.1 | - | 13 - 14 | 1137 | 79 | |
AK296837.1 | - | 13 - 14 | 1214 | 79 | |
AK299229.1 | - | 9 - 10 | 875 | 79 | |
AK299769.1 | - | 12 - 13 | 1157 | 79 | |
AK304558.1 | - | 13 - 14 | 1190 | 79 | |
AK315932.1 | - | 9 - 10 | 880 | 79 | |
AL096748.1 | - | 1 - 2 | 704 | 79 | |
BC032661.1 | - | 15 - 16 | 1547 | 79 | |
BC041699.1 | - | 15 - 16 | 1362 | 79 |