Assay Design



Gene Transcripts

Gene Symbol
GSTO1
Entrez Gene ID
9446
Gene Name
glutathione S-transferase omega 1
Gene Aliases
GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
Chromosome Location
Chr.10: 104254194 - 104267464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191003.1 NP_001177932.1 1 - 2 305 86
GenBank mRNA DB501888.1 - 1 - 2 336 86

View Full Product Details