Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001269039.2 | NP_001255968.1 | 4 - 5 | 1368 | 120 |
NM_001324336.1 | NP_001311265.1 | 5 - 6 | 1463 | 120 | |
NM_001324337.1 | NP_001311266.1 | 6 - 7 | 1691 | 120 | |
NM_007373.3 | NP_031399.2 | 5 - 6 | 1506 | 120 | |
NR_136749.1 | 4 - 5 | 569 | 120 | ||
XM_017016702.1 | XP_016872191.1 | 5 - 6 | 1411 | 120 | |
XM_017016703.1 | XP_016872192.1 | 5 - 6 | 7228 | 120 | |
XM_017016704.1 | XP_016872193.1 | 5 - 6 | 1410 | 120 | |
GenBank mRNA | AB020669.1 | - | 5 - 6 | 1434 | 120 |
AF054828.1 | - | 5 - 6 | 1428 | 120 | |
AF068920.1 | - | 4 - 5 | 1157 | 120 | |
AK090820.1 | - | 3 - 4 | 527 | 120 | |
AK292833.1 | - | 6 - 7 | 1632 | 120 | |
AK293271.1 | - | 4 - 5 | 508 | 120 | |
BC044752.1 | - | 5 - 6 | 1442 | 120 | |
BC050445.1 | - | 4 - 5 | 1361 | 120 | |
BX649055.1 | - | 6 - 7 | 1274 | 120 |