Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016034.4 | NP_057118.1 | 3 - 4 | 377 | 80 |
NR_051969.1 | 3 - 4 | 580 | 80 | ||
NR_051970.1 | 3 - 4 | 522 | 80 | ||
XM_006717136.3 | XP_006717199.1 | 4 - 5 | 501 | 80 | |
GenBank mRNA | AB055388.1 | - | 3 - 4 | 301 | 80 |
AF151849.1 | - | 3 - 4 | 301 | 80 | |
BC008017.1 | - | 3 - 4 | 313 | 80 | |
BC013108.2 | - | 3 - 4 | 321 | 80 | |
BE264885.1 | - | 3 - 4 | 496 | 80 | |
BU169388.1 | - | 3 - 4 | 438 | 80 | |
BX362434.2 | - | 3 - 4 | 371 | 80 |