Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001635.3 | NP_001626.1 | 19 - 20 | 2098 | 89 |
NM_139316.2 | NP_647477.1 | 18 - 19 | 1972 | 89 | |
XM_006715689.3 | XP_006715752.1 | 18 - 19 | 3635 | 89 | |
XM_006715690.3 | XP_006715753.1 | 18 - 19 | 2417 | 89 | |
XM_011515271.2 | XP_011513573.1 | 19 - 20 | 2543 | 89 | |
XM_017011995.1 | XP_016867484.1 | 19 - 20 | 3761 | 89 | |
XM_017011996.1 | XP_016867485.1 | 18 - 19 | 2723 | 89 | |
GenBank mRNA | AF034996.1 | - | 18 - 19 | 1825 | 89 |
AF498092.1 | - | 7 - 8 | 1153 | 89 | |
AF498096.1 | - | 10 - 11 | 1548 | 89 | |
AK291447.1 | - | 19 - 20 | 1902 | 89 | |
BC034376.1 | - | 19 - 20 | 1889 | 89 | |
U07616.1 | - | 19 - 20 | 1992 | 89 | |
X81438.1 | - | 19 - 20 | 1956 | 89 |