Assay Design



Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr.7: 38383694 - 38631567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001635.3 NP_001626.1 2 - 3 368 92
NM_139316.2 NP_647477.1 2 - 3 368 92
XM_006715689.3 XP_006715752.1 2 - 3 315 92
XM_006715690.3 XP_006715753.1 2 - 3 315 92
XM_006715691.3 XP_006715754.1 2 - 3 315 92
XM_011515271.2 XP_011513573.1 2 - 3 315 92
XM_017011995.1 XP_016867484.1 2 - 3 315 92
XM_017011996.1 XP_016867485.1 2 - 3 315 92
GenBank mRNA AF034996.1 - 2 - 3 221 92
AF498097.1 - 2 - 3 152 92
AK291447.1 - 2 - 3 172 92
BC034376.1 - 2 - 3 159 92
DA125756.1 - 2 - 3 368 92
U07616.1 - 2 - 3 262 92
X81438.1 - 2 - 3 226 92

View Full Product Details