Assay Design



Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
79639
Gene Name
transmembrane protein 53
Gene Aliases
NET4
Chromosome Location
Chr.1: 44653247 - 44674608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011542138.2 XP_011540440.1 1 - 2 496 86
GenBank mRNA AK310016.1 - 1 - 2 311 86

View Full Product Details