Assay Design
Gene Transcripts
- Gene Symbol
- TMEM53
- Entrez Gene ID
-
79639
- Gene Name
- transmembrane protein 53
- Gene Aliases
-
NET4
- Chromosome Location
-
Chr.1: 44653247 - 44674608 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BM806701.1 |
- |
3 - 4 |
284 |
68 |
View Full Product Details