Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001118887.1 | NP_001112359.1 | 3 - 4 | 902 | 95 |
NM_001118888.1 | NP_001112360.1 | 2 - 3 | 746 | 95 | |
NM_001147.2 | NP_001138.1 | 3 - 4 | 902 | 95 | |
XM_017013318.1 | XP_016868807.1 | 2 - 3 | 903 | 95 | |
GenBank mRNA | AB009865.1 | - | 3 - 4 | 601 | 95 |
AF004327.1 | - | 3 - 4 | 922 | 95 | |
AF187858.1 | - | 2 - 3 | 437 | 95 | |
AF218015.1 | - | 3 - 4 | 655 | 95 | |
AK075219.1 | - | 3 - 4 | 884 | 95 | |
AK225698.1 | - | 3 - 4 | 884 | 95 | |
AK290070.1 | - | 3 - 4 | 884 | 95 | |
AK310171.1 | - | 3 - 4 | 884 | 95 | |
AK312541.1 | - | 3 - 4 | 884 | 95 | |
BC126200.1 | - | 3 - 4 | 630 | 95 | |
BC126202.1 | - | 3 - 4 | 630 | 95 | |
BC143902.1 | - | 3 - 4 | 630 | 95 |