Assay Design



Gene Transcripts

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
Chromosome Location
Chr.3: 134355345 - 134375417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278683.1 NP_001265612.1 3 - 4 1334 90
NM_001278685.1 NP_001265614.1 3 - 4 1348 90
NM_016201.3 NP_057285.3 3 - 4 1348 90
XM_006713654.2 XP_006713717.1 3 - 4 1366 90
XM_011512881.1 XP_011511183.1 3 - 4 1205 90
XM_017006580.1 XP_016862069.1 3 - 4 1205 90
XM_017006581.1 XP_016862070.1 3 - 4 1386 90
XM_017006582.1 XP_016862071.1 3 - 4 1213 90
GenBank mRNA AB023206.1 - 3 - 4 1281 90
AF175966.1 - 2 - 3 415 90
AK001343.1 - 3 - 4 1211 90
AK223598.1 - 3 - 4 1206 90
AK291616.1 - 3 - 4 1292 90
AK299270.1 - 3 - 4 1334 90
BC011454.1 - 1 - 2 314 90

View Full Product Details