Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142536.1 | NP_001136008.1 | 2 - 3 | 561 | 60 |
NM_001142537.1 | NP_001136009.1 | 1 - 2 | 651 | 60 | |
NM_001142538.1 | NP_001136010.1 | 1 - 2 | 651 | 60 | |
NM_033101.3 | NP_149092.2 | 2 - 3 | 561 | 60 | |
GenBank mRNA | AF222695.1 | - | 1 - 2 | 131 | 60 |
AF244974.1 | - | 2 - 3 | 530 | 60 | |
AF244975.1 | - | 1 - 2 | 651 | 60 | |
AF244976.1 | - | 2 - 3 | 530 | 60 | |
AF244977.1 | - | 1 - 2 | 651 | 60 | |
AF310686.1 | - | 2 - 3 | 175 | 60 | |
AK313851.1 | - | 2 - 3 | 561 | 60 | |
BC028222.1 | - | 2 - 3 | 511 | 60 |