Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323352.1 | NP_001310281.1 | 2 - 3 | 470 | 69 |
NM_001323360.1 | NP_001310289.1 | 2 - 3 | 470 | 69 | |
NM_001323362.1 | NP_001310291.1 | 2 - 3 | 470 | 69 | |
NM_001323364.1 | NP_001310293.1 | 2 - 3 | 481 | 69 | |
NM_001323365.1 | NP_001310294.1 | 2 - 3 | 309 | 69 | |
NM_001323366.1 | NP_001310295.1 | 2 - 3 | 309 | 69 | |
NM_006100.3 | NP_006091.1 | 2 - 3 | 481 | 69 | |
GenBank mRNA | AK315111.1 | - | 2 - 3 | 472 | 69 |
BC023312.1 | - | 2 - 3 | 457 | 69 | |
DB036799.1 | - | 2 - 3 | 471 | 69 |