Assay Design



Gene Transcripts

Gene Symbol
ACSM1
Entrez Gene ID
116285
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
BUCS1, MACS1
Chromosome Location
Chr.16: 20623235 - 20697788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318890.1 NP_001305819.1 5 - 6 959 81
NM_052956.2 NP_443188.2 4 - 5 823 81
XM_006721016.2 XP_006721079.1 4 - 5 802 81
XM_006721017.2 XP_006721080.1 4 - 5 802 81
XM_006721018.2 XP_006721081.1 2 - 3 181 81
XM_011545729.2 XP_011544031.1 4 - 5 802 81
XM_011545730.2 XP_011544032.1 4 - 5 802 81
XM_011545731.2 XP_011544033.1 2 - 3 998 81
XM_011545732.2 XP_011544034.1 3 - 4 240 81
XM_017022914.1 XP_016878403.1 4 - 5 802 81
GenBank mRNA AB059429.1 - 4 - 5 823 81
AK092574.1 - 2 - 3 181 81
AK314282.1 - 5 - 6 915 81
BC125178.1 - 4 - 5 823 81

View Full Product Details