Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017392.4 | NP_001017392.2 | 2 - 3 | 274 | 82 |
NM_001321697.1 | NP_001308626.1 | 2 - 3 | 430 | 82 | |
NM_001321698.1 | NP_001308627.1 | 2 - 3 | 222 | 82 | |
NM_001321699.1 | NP_001308628.1 | 2 - 3 | 222 | 82 | |
NM_014884.4 | NP_055699.2 | 2 - 3 | 274 | 82 | |
XM_017026137.1 | XP_016881626.1 | 2 - 3 | 430 | 82 | |
GenBank mRNA | AB002363.2 | - | 2 - 3 | 426 | 82 |
AF518874.1 | - | 2 - 3 | 263 | 82 | |
AK074842.1 | - | 2 - 3 | 199 | 82 | |
AK291275.1 | - | 2 - 3 | 241 | 82 | |
AK299860.1 | - | 1 - 2 | 87 | 82 | |
AL832488.1 | - | 2 - 3 | 194 | 82 | |
BC020586.1 | - | 2 - 3 | 194 | 82 | |
BX647813.1 | - | 2 - 3 | 203 | 82 | |
DA217559.1 | - | 2 - 3 | 82 | ||
DA770920.1 | - | 2 - 3 | 417 | 82 | |
DC335817.1 | - | 2 - 3 | 274 | 82 | |
DC402794.1 | - | 2 - 3 | 222 | 82 |