Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017392.4 | NP_001017392.2 | 9 - 10 | 3279 | 121 |
NM_001321697.1 | NP_001308626.1 | 9 - 10 | 3435 | 121 | |
NM_001321698.1 | NP_001308627.1 | 9 - 10 | 3227 | 121 | |
NM_001321699.1 | NP_001308628.1 | 9 - 10 | 3227 | 121 | |
NM_014884.4 | NP_055699.2 | 9 - 10 | 3279 | 121 | |
XM_017026137.1 | XP_016881626.1 | 9 - 10 | 3435 | 121 | |
GenBank mRNA | AB002363.2 | - | 9 - 10 | 3431 | 121 |
AF518874.1 | - | 9 - 10 | 3268 | 121 | |
AK291275.1 | - | 9 - 10 | 3246 | 121 | |
AL832488.1 | - | 10 - 11 | 3043 | 121 | |
BX647813.1 | - | 9 - 10 | 3208 | 121 |