Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015110.3 | NP_055925.2 | 9 - 10 | 1427 | 120 |
XM_005251837.1 | XP_005251894.1 | 9 - 10 | 1427 | 120 | |
XM_005251838.1 | XP_005251895.1 | 8 - 9 | 1355 | 120 | |
XM_005251839.1 | XP_005251896.1 | 9 - 10 | 1427 | 120 | |
XM_017014507.1 | XP_016869996.1 | 9 - 10 | 1427 | 120 | |
XM_017014508.1 | XP_016869997.1 | 9 - 10 | 1427 | 120 | |
XM_017014509.1 | XP_016869998.1 | 10 - 11 | 1561 | 120 | |
XM_017014510.1 | XP_016869999.1 | 2 - 3 | 391 | 120 | |
GenBank mRNA | AB011166.2 | - | 9 - 10 | 1368 | 120 |
AJ310550.1 | - | 9 - 10 | 1310 | 120 | |
AK055825.1 | - | 9 - 10 | 1339 | 120 | |
AK304133.1 | - | 9 - 10 | 1427 | 120 | |
BC017666.1 | - | 9 - 10 | 1386 | 120 | |
BC038225.1 | - | 9 - 10 | 1410 | 120 |