Assay Design
Gene Transcripts
- Gene Symbol
- BCL2
- Entrez Gene ID
-
596
- Gene Name
- BCL2, apoptosis regulator
- Gene Aliases
-
Bcl-2, PPP1R50
- Chromosome Location
-
Chr.18: 63123346 - 63319778 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M13995.1 |
- |
1 - 1 |
854 |
68 |
View Full Product Details