Assay Design
Gene Transcripts
- Gene Symbol
- TTC1
- Entrez Gene ID
-
7265
- Gene Name
- tetratricopeptide repeat domain 1
- Gene Aliases
-
TPR1
- Chromosome Location
-
Chr.5: 160009100 - 160065545 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003314.2 |
NP_003305.1 |
1 - 2 |
91 |
100 |
GenBank mRNA |
BP245572.1 |
- |
1 - 2 |
91 |
100 |
View Full Product Details