Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282500.1 | NP_001269429.1 | 4 - 5 | 619 | 88 |
NM_003314.2 | NP_003305.1 | 4 - 5 | 622 | 88 | |
GenBank mRNA | AK222641.1 | - | 4 - 5 | 573 | 88 |
AK222984.1 | - | 4 - 5 | 567 | 88 | |
AK315262.1 | - | 4 - 5 | 577 | 88 | |
BC000942.3 | - | 3 - 4 | 527 | 88 | |
DN989835.1 | - | 4 - 5 | 575 | 88 | |
U46570.1 | - | 4 - 5 | 549 | 88 |