Assay Design



Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr.16: 74873568 - 74985146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030581.3 NP_085058.3 24 - 25 2655 111
XM_005256146.3 XP_005256203.1 25 - 26 2716 111
XM_011523331.2 XP_011521633.1 23 - 24 2476 111
XM_011523332.2 XP_011521634.1 19 - 20 2164 111
XM_017023667.1 XP_016879156.1 23 - 24 2516 111
XM_017023668.1 XP_016879157.1 22 - 23 2292 111
XM_017023669.1 XP_016879158.1 23 - 24 2364 111
XM_017023670.1 XP_016879159.1 22 - 23 2397 111
GenBank mRNA AB067510.1 - 19 - 20 2128 111
AF370390.1 - 12 - 13 1587 111
AK022332.1 - 7 - 8 1087 111
AK091316.1 - 12 - 13 1435 111
AL832078.1 - 9 - 10 1063 111
BC014887.2 - 18 - 19 2047 111

View Full Product Details