Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324171.1 | NP_001311100.1 | 9 - 10 | 856 | 68 |
NM_001324172.1 | NP_001311101.1 | 9 - 10 | 856 | 68 | |
NM_030581.3 | NP_085058.3 | 9 - 10 | 829 | 68 | |
XM_005256146.3 | XP_005256203.1 | 9 - 10 | 833 | 68 | |
XM_011523331.2 | XP_011521633.1 | 8 - 9 | 673 | 68 | |
XM_011523332.2 | XP_011521634.1 | 3 - 4 | 256 | 68 | |
XM_017023667.1 | XP_016879156.1 | 8 - 9 | 690 | 68 | |
XM_017023668.1 | XP_016879157.1 | 7 - 8 | 466 | 68 | |
XM_017023669.1 | XP_016879158.1 | 8 - 9 | 561 | 68 | |
XM_017023670.1 | XP_016879159.1 | 7 - 8 | 569 | 68 | |
GenBank mRNA | AB067510.1 | - | 4 - 5 | 302 | 68 |
AK302557.1 | - | 9 - 10 | 897 | 68 | |
AL562571.3 | - | 5 - 6 | 68 | ||
BC004519.2 | - | 9 - 10 | 793 | 68 | |
BC014887.2 | - | 3 - 4 | 221 | 68 |