Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324171.1 | NP_001311100.1 | 14 - 15 | 1512 | 59 |
NM_030581.3 | NP_085058.3 | 14 - 15 | 1485 | 59 | |
XM_005256146.3 | XP_005256203.1 | 14 - 15 | 1489 | 59 | |
XM_011523331.2 | XP_011521633.1 | 12 - 13 | 1249 | 59 | |
XM_011523332.2 | XP_011521634.1 | 8 - 9 | 937 | 59 | |
XM_017023667.1 | XP_016879156.1 | 13 - 14 | 1346 | 59 | |
XM_017023668.1 | XP_016879157.1 | 12 - 13 | 1122 | 59 | |
XM_017023669.1 | XP_016879158.1 | 12 - 13 | 1137 | 59 | |
XM_017023670.1 | XP_016879159.1 | 11 - 12 | 1170 | 59 | |
GenBank mRNA | AB067510.1 | - | 9 - 10 | 958 | 59 |
AF370390.1 | - | 2 - 3 | 417 | 59 | |
AK091316.1 | - | 2 - 3 | 265 | 59 | |
BC004519.2 | - | 14 - 15 | 1449 | 59 | |
BC014887.2 | - | 8 - 9 | 877 | 59 |