Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001155.4 | NP_001146.2 | 24 - 25 | 2067 | 76 |
NM_001193544.1 | NP_001180473.1 | 23 - 24 | 1944 | 76 | |
XM_005268432.4 | XP_005268489.1 | 23 - 24 | 2049 | 76 | |
GenBank mRNA | AK126836.1 | - | - | 2052 | 76 |
AK130077.1 | - | 13 - 14 | 908 | 76 | |
AK290672.1 | - | 24 - 25 | 1932 | 76 | |
AK298582.1 | - | 23 - 24 | 1724 | 76 | |
BC017046.1 | - | 24 - 25 | 1888 | 76 | |
D00510.1 | - | 23 - 24 | 1839 | 76 | |
J03578.1 | - | 24 - 25 | 1895 | 76 | |
KU177903.1 | - | 23 - 24 | 1839 | 76 | |
KU177904.1 | - | 22 - 23 | 1821 | 76 | |
KU177905.1 | - | 15 - 16 | 1070 | 76 |