Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322123.1 | NP_001309052.1 | 16 - 17 | 1660 | 65 |
NM_001322124.1 | NP_001309053.1 | 16 - 17 | 1529 | 65 | |
NM_001322127.1 | NP_001309056.1 | 15 - 16 | 1655 | 65 | |
NM_144709.3 | NP_653310.2 | 16 - 17 | 1717 | 65 | |
XM_011532568.2 | XP_011530870.1 | 17 - 18 | 1788 | 65 | |
XM_011532570.1 | XP_011530872.1 | 16 - 17 | 1495 | 65 | |
XM_011532571.1 | XP_011530873.1 | 16 - 17 | 1669 | 65 | |
XM_011532572.1 | XP_011530874.1 | 16 - 17 | 1589 | 65 | |
XM_011532573.2 | XP_011530875.1 | 16 - 17 | 1549 | 65 | |
XM_011532574.2 | XP_011530876.1 | 16 - 17 | 1610 | 65 | |
XM_011532576.2 | XP_011530878.1 | 16 - 17 | 1669 | 65 | |
XM_011532578.1 | XP_011530880.1 | 16 - 17 | 1647 | 65 | |
XM_017003428.1 | XP_016858917.1 | 15 - 16 | 1486 | 65 | |
XM_017003429.1 | XP_016858918.1 | 15 - 16 | 1282 | 65 | |
GenBank mRNA | AK056874.1 | - | 16 - 17 | 1531 | 65 |
AK291729.1 | - | 16 - 17 | 1505 | 65 | |
AL832208.1 | - | 14 - 15 | 4056 | 65 | |
BC101680.1 | - | 16 - 17 | 1509 | 65 | |
BC101706.1 | - | 16 - 17 | 1509 | 65 | |
BC143480.1 | - | 15 - 16 | 1447 | 65 |