Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322123.1 | NP_001309052.1 | 17 - 18 | 1762 | 71 |
NM_001322124.1 | NP_001309053.1 | 17 - 18 | 1631 | 71 | |
NM_001322127.1 | NP_001309056.1 | 16 - 17 | 1757 | 71 | |
NM_144709.3 | NP_653310.2 | 17 - 18 | 1819 | 71 | |
XM_011532568.2 | XP_011530870.1 | 18 - 19 | 1890 | 71 | |
XM_011532570.1 | XP_011530872.1 | 17 - 18 | 1597 | 71 | |
XM_011532571.1 | XP_011530873.1 | 17 - 18 | 1771 | 71 | |
XM_011532572.1 | XP_011530874.1 | 17 - 18 | 1691 | 71 | |
XM_011532573.2 | XP_011530875.1 | 17 - 18 | 1651 | 71 | |
XM_011532574.2 | XP_011530876.1 | 17 - 18 | 1712 | 71 | |
XM_011532576.2 | XP_011530878.1 | 17 - 18 | 1771 | 71 | |
XM_011532578.1 | XP_011530880.1 | 17 - 18 | 1749 | 71 | |
XM_017003428.1 | XP_016858917.1 | 16 - 17 | 1588 | 71 | |
XM_017003429.1 | XP_016858918.1 | 16 - 17 | 1384 | 71 | |
GenBank mRNA | AK056874.1 | - | 17 - 18 | 1633 | 71 |
AK291729.1 | - | 17 - 18 | 1607 | 71 | |
AL832208.1 | - | 15 - 16 | 4158 | 71 | |
BC101680.1 | - | 17 - 18 | 1611 | 71 | |
BC101706.1 | - | 17 - 18 | 1611 | 71 | |
BC143480.1 | - | 16 - 17 | 1549 | 71 |