Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040439.1 | NP_001035529.1 | 1 - 2 | 429 | 76 |
NM_001318852.1 | NP_001305781.1 | 1 - 2 | 466 | 76 | |
NM_015133.4 | NP_055948.2 | 1 - 2 | 466 | 76 | |
XM_005255190.2 | XP_005255247.1 | 1 - 2 | 466 | 76 | |
XM_006720869.2 | XP_006720932.1 | 1 - 2 | 269 | 76 | |
XM_011522429.2 | XP_011520731.1 | 1 - 2 | 466 | 76 | |
XM_011522430.2 | XP_011520732.1 | 1 - 2 | 466 | 76 | |
XM_011522431.2 | XP_011520733.1 | 1 - 2 | 466 | 76 | |
XM_011522432.2 | XP_011520734.1 | 1 - 2 | 466 | 76 | |
XM_017023079.1 | XP_016878568.1 | 1 - 2 | 466 | 76 | |
XM_017023080.1 | XP_016878569.1 | 1 - 2 | 466 | 76 | |
GenBank mRNA | AB028989.2 | - | 1 - 2 | 429 | 76 |
AB621811.1 | - | 1 - 2 | 454 | 76 | |
AK024437.1 | - | 1 - 2 | 408 | 76 | |
BC137123.1 | - | 1 - 2 | 381 | 76 | |
BC137124.1 | - | 1 - 2 | 381 | 76 | |
BC144486.1 | - | 1 - 2 | 381 | 76 | |
BC150266.1 | - | 1 - 2 | 429 | 76 | |
BY795428.2 | - | 1 - 2 | 189 | 76 | |
CN405914.1 | - | 1 - 2 | 458 | 76 | |
CR749696.1 | - | 1 - 2 | 269 | 76 | |
DA135446.1 | - | 1 - 2 | 466 | 76 |