Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130926.2 | NP_001124398.1 | 1 - 2 | 175 | 131 |
NM_001130927.2 | NP_001124399.1 | 1 - 2 | 175 | 131 | |
XM_011521576.2 | XP_011519878.1 | 1 - 2 | 111 | 131 | |
XM_011521583.2 | XP_011519885.1 | 1 - 2 | 111 | 131 | |
XM_017022190.1 | XP_016877679.1 | 1 - 2 | 111 | 131 | |
XM_017022192.1 | XP_016877681.1 | 1 - 2 | 100 | 131 | |
XM_017022194.1 | XP_016877683.1 | 1 - 2 | 111 | 131 | |
XM_017022201.1 | XP_016877690.1 | 1 - 2 | 108 | 131 | |
GenBank mRNA | AK294207.1 | - | 1 - 2 | 113 | 131 |