Assay Design



Gene Transcripts

Gene Symbol
MEF2A
Entrez Gene ID
4205
Gene Name
myocyte enhancer factor 2A
Gene Aliases
ADCAD1, RSRFC4, RSRFC9, mef2
Chromosome Location
Chr.15: 99565455 - 99716488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130926.2 NP_001124398.1 6 - 7 986 88
NM_001130927.2 NP_001124399.1 5 - 6 782 88
NM_001130928.2 NP_001124400.1 4 - 5 605 88
NM_001171894.2 NP_001165365.1 8 - 9 1276 88
NM_001319206.1 NP_001306135.1 7 - 8 1068 88
NM_005587.3 NP_005578.2 7 - 8 1074 88
XM_005254914.2 XP_005254971.1 7 - 8 1051 88
XM_005254915.2 XP_005254972.1 7 - 8 1296 88
XM_011521571.2 XP_011519873.1 7 - 8 1051 88
XM_011521572.2 XP_011519874.1 7 - 8 996 88
XM_011521573.2 XP_011519875.1 7 - 8 1026 88
XM_011521576.2 XP_011519878.1 6 - 7 940 88
XM_011521577.2 XP_011519879.1 6 - 7 874 88
XM_011521578.2 XP_011519880.1 6 - 7 1533 88
XM_011521579.2 XP_011519881.1 6 - 7 950 88
XM_011521581.2 XP_011519883.1 7 - 8 1302 88
XM_011521582.2 XP_011519884.1 8 - 9 1507 88
XM_011521583.2 XP_011519885.1 6 - 7 922 88
XM_011521585.2 XP_011519887.1 6 - 7 1110 88
XM_011521586.2 XP_011519888.1 6 - 7 1092 88
XM_011521587.2 XP_011519889.1 6 - 7 1092 88
XM_011521590.2 XP_011519892.1 7 - 8 1051 88
XM_017022190.1 XP_016877679.1 6 - 7 928 88
XM_017022191.1 XP_016877680.1 6 - 7 920 88
XM_017022192.1 XP_016877681.1 6 - 7 929 88
XM_017022193.1 XP_016877682.1 7 - 8 1112 88
XM_017022194.1 XP_016877683.1 6 - 7 928 88
XM_017022195.1 XP_016877684.1 6 - 7 785 88
XM_017022196.1 XP_016877685.1 8 - 9 1440 88
XM_017022197.1 XP_016877686.1 7 - 8 3219 88
XM_017022198.1 XP_016877687.1 8 - 9 1428 88
XM_017022199.1 XP_016877688.1 6 - 7 1729 88
XM_017022200.1 XP_016877689.1 7 - 8 1499 88
XM_017022201.1 XP_016877690.1 6 - 7 1122 88
XM_017022202.1 XP_016877691.1 8 - 9 1422 88
GenBank mRNA AB208985.1 - 8 - 9 1253 88
AK291835.1 - 6 - 7 896 88
AK294207.1 - 5 - 6 720 88
AL831995.1 - 7 - 8 929 88
BC013437.2 - 7 - 8 992 88
BC053871.2 - 4 - 5 577 88
KU178181.1 - 5 - 6 673 88
KU178182.1 - 5 - 6 673 88
KU178183.1 - 5 - 6 667 88
KU178184.1 - 5 - 6 667 88
KU178185.1 - 4 - 5 463 88
X63381.1 - 6 - 7 901 88
X68505.1 - 7 - 8 1087 88
Y16312.1 - 6 - 7 895 88

View Full Product Details