Assay Design



Gene Transcripts

Gene Symbol
MEF2A
Entrez Gene ID
4205
Gene Name
myocyte enhancer factor 2A
Gene Aliases
ADCAD1, RSRFC4, RSRFC9, mef2
Chromosome Location
Chr.15: 99565455 - 99716488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005587.3 NP_005578.2 4 - 5 658 66
XM_005254914.2 XP_005254971.1 4 - 5 623 66
XM_005254916.3 XP_005254973.1 4 - 5 623 66
XM_011521571.2 XP_011519873.1 4 - 5 623 66
XM_011521572.2 XP_011519874.1 4 - 5 568 66
XM_011521573.2 XP_011519875.1 4 - 5 598 66
XM_011521576.2 XP_011519878.1 3 - 4 512 66
XM_011521577.2 XP_011519879.1 3 - 4 446 66
XM_011521578.2 XP_011519880.1 3 - 4 1105 66
XM_011521579.2 XP_011519881.1 3 - 4 522 66
XM_011521581.2 XP_011519883.1 4 - 5 886 66
XM_011521582.2 XP_011519884.1 5 - 6 1091 66
XM_011521590.2 XP_011519892.1 4 - 5 623 66
XM_017022190.1 XP_016877679.1 3 - 4 512 66
XM_017022191.1 XP_016877680.1 3 - 4 504 66
XM_017022192.1 XP_016877681.1 3 - 4 501 66
XM_017022193.1 XP_016877682.1 4 - 5 696 66
XM_017022194.1 XP_016877683.1 3 - 4 512 66
XM_017022203.1 XP_016877692.1 4 - 5 886 66
GenBank mRNA BC013437.2 - 4 - 5 576 66
KU178181.1 - 2 - 3 257 66
KU178182.1 - 2 - 3 257 66
X63381.1 - 3 - 4 485 66
X68505.1 - 4 - 5 671 66

View Full Product Details