Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152501.4 | NP_689714.2 | 2 - 3 | 516 | 133 |
NM_198929.4 | NP_945147.1 | 2 - 3 | 516 | 133 | |
XM_005244930.1 | XP_005244987.1 | 2 - 3 | 516 | 133 | |
XM_011509242.2 | XP_011507544.1 | 2 - 3 | 516 | 133 | |
XM_011509243.2 | XP_011507545.1 | 2 - 3 | 516 | 133 | |
XM_017000463.1 | XP_016855952.1 | 2 - 3 | 516 | 133 | |
GenBank mRNA | AK308350.1 | - | 2 - 3 | 495 | 133 |
AY185344.1 | - | 2 - 3 | 503 | 133 | |
AY185346.1 | - | 2 - 3 | 503 | 133 | |
BC020822.1 | - | 2 - 3 | 495 | 133 | |
BC090944.1 | - | 2 - 3 | 498 | 133 |