Assay Design
Gene Transcripts
- Gene Symbol
- MYL12B
- Entrez Gene ID
-
103910
- Gene Name
- myosin light chain 12B
- Gene Aliases
-
MLC-B, MRLC2
- Chromosome Location
-
Chr.18: 3262113 - 3278284 on Build GRCh38
- Gene Symbol
- LOC104968399
- Entrez Gene ID
-
104968399
- Gene Name
- uncharacterized LOC104968399
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 3255436 - 3262359 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL550084.3 |
- |
3 - 4 |
|
54 |
HY167137.1 |
- |
1 - 2 |
|
54 |
View Full Product Details