Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301132.1 | NP_001288061.1 | 6 - 7 | 102 | |
NM_014952.4 | NP_055767.3 | 6 - 7 | 102 | ||
NR_125372.1 | 4 - 5 | 102 | |||
XM_011521366.2 | XP_011519668.2 | 6 - 7 | 102 | ||
XM_011521367.2 | XP_011519669.2 | 6 - 7 | 102 | ||
XM_011521368.2 | XP_011519670.2 | 6 - 7 | 102 | ||
XM_011521369.2 | XP_011519671.1 | 6 - 7 | 102 | ||
GenBank mRNA | AB023162.1 | - | 6 - 7 | 102 | |
AL833923.1 | - | 5 - 6 | 102 | ||
BC022782.2 | - | 6 - 7 | 102 | ||
HY353002.1 | - | 1 - 2 | 94 | 102 |