Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199196.1 | NP_001186125.1 | 7 - 8 | 1588 | 85 |
NM_033415.3 | NP_219483.1 | 6 - 7 | 1480 | 85 | |
XM_005260157.3 | XP_005260214.1 | 6 - 7 | 1531 | 85 | |
XM_017027484.1 | XP_016882973.1 | 6 - 7 | 1455 | 85 | |
XM_017027485.1 | XP_016882974.1 | 8 - 9 | 1712 | 85 | |
GenBank mRNA | AK295488.1 | - | 6 - 7 | 1254 | 85 |
AK303914.1 | - | 4 - 5 | 1168 | 85 | |
BC003700.1 | - | 2 - 3 | 226 | 85 | |
BC056161.1 | - | 8 - 9 | 1210 | 85 | |
BC067125.1 | - | 7 - 8 | 1357 | 85 | |
CR749678.1 | - | 3 - 4 | 821 | 85 |