Assay Design



Gene Transcripts

Gene Symbol
TSPAN3
Entrez Gene ID
10099
Gene Name
tetraspanin 3
Gene Aliases
TM4-A, TM4SF8, TSPAN-3
Chromosome Location
Chr.15: 77044017 - 77071228 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168412.1 NP_001161884.1 2 - 3 413 85
NM_005724.5 NP_005715.1 3 - 4 605 85
XM_017021857.1 XP_016877346.1 3 - 4 214 85
GenBank mRNA AF054840.1 - 3 - 4 331 85
AF133423.1 - 3 - 4 405 85
AK001305.1 - 3 - 4 451 85
AK001310.1 - 3 - 4 437 85
AK001326.1 - 3 - 4 548 85
AK075336.1 - 3 - 4 486 85
AK293677.1 - 4 - 5 394 85
AK298157.1 - 2 - 3 294 85
BC000704.2 - 3 - 4 506 85
BC004280.1 - 3 - 4 467 85
BC009248.2 - 3 - 4 470 85
BC011206.1 - 3 - 4 499 85

View Full Product Details