Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168355.2 | NP_001161827.1 | 3 - 4 | 522 | 136 |
NM_001323504.1 | NP_001310433.1 | 3 - 4 | 642 | 136 | |
NM_001323505.1 | NP_001310434.1 | 3 - 4 | 522 | 136 | |
NM_001323506.1 | NP_001310435.1 | 3 - 4 | 522 | 136 | |
NM_001323507.1 | NP_001310436.1 | 3 - 4 | 522 | 136 | |
NM_003999.2 | NP_003990.1 | 3 - 4 | 642 | 136 | |
XM_005248384.1 | XP_005248441.1 | 3 - 4 | 629 | 136 | |
XM_005248386.2 | XP_005248443.1 | 3 - 4 | 629 | 136 | |
XM_005248387.2 | XP_005248444.1 | 3 - 4 | 629 | 136 | |
XM_011514161.2 | XP_011512463.1 | 3 - 4 | 629 | 136 | |
XM_017010019.1 | XP_016865508.1 | 3 - 4 | 629 | 136 | |
GenBank mRNA | AK304016.1 | - | 3 - 4 | 596 | 136 |
BC010943.1 | - | 3 - 4 | 372 | 136 | |
BC063468.1 | - | 3 - 4 | 294 | 136 | |
BC125209.1 | - | 3 - 4 | 412 | 136 | |
BC125210.1 | - | 3 - 4 | 412 | 136 | |
DR006275.1 | - | 3 - 4 | 649 | 136 | |
U60805.1 | - | 3 - 4 | 607 | 136 |