Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005530.2 | NP_005521.1 | 9 - 10 | 891 | 88 |
XM_005254336.2 | XP_005254393.1 | 8 - 9 | 827 | 88 | |
XM_017022118.1 | XP_016877607.1 | 8 - 9 | 906 | 88 | |
XM_017022119.1 | XP_016877608.1 | 8 - 9 | 996 | 88 | |
XM_017022120.1 | XP_016877609.1 | 7 - 8 | 791 | 88 | |
GenBank mRNA | AK222973.1 | - | 9 - 10 | 875 | 88 |
AK296002.1 | - | 9 - 10 | 866 | 88 | |
AK299970.1 | - | 9 - 10 | 803 | 88 | |
AK315963.1 | - | 8 - 9 | 775 | 88 | |
AK316051.1 | - | 8 - 9 | 763 | 88 | |
AL110096.1 | - | 5 - 6 | 538 | 88 | |
AL442090.1 | - | 6 - 7 | 813 | 88 | |
BC021967.2 | - | 9 - 10 | 884 | 88 | |
BC036227.1 | - | 9 - 10 | 892 | 88 | |
CR749548.1 | - | 9 - 10 | 1555 | 88 | |
U07681.1 | - | 9 - 10 | 869 | 88 |