Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000211.4 | NP_000202.3 | 13 - 14 | 2104 | 97 |
NM_001127491.2 | NP_001120963.2 | 13 - 14 | 2094 | 97 | |
NM_001303238.1 | NP_001290167.1 | 13 - 14 | 2059 | 97 | |
XM_006724001.1 | XP_006724064.1 | 13 - 14 | 2121 | 97 | |
XM_017028341.1 | XP_016883830.1 | 13 - 14 | 2116 | 97 | |
GenBank mRNA | AB208909.1 | - | 12 - 13 | 3927 | 97 |
AK095992.1 | - | 13 - 14 | 2059 | 97 | |
AK097216.1 | - | 4 - 5 | 1271 | 97 | |
AK128344.1 | - | 13 - 14 | 1929 | 97 | |
AK222505.1 | - | 13 - 14 | 1961 | 97 | |
AK303488.1 | - | 13 - 14 | 1972 | 97 | |
BC005861.2 | - | 12 - 13 | 1915 | 97 | |
BC021077.1 | - | 12 - 13 | 1838 | 97 | |
L78790.1 | - | 12 - 13 | 1751 | 97 | |
M15395.1 | - | 13 - 14 | 1943 | 97 | |
Y00057.1 | - | 12 - 13 | 1847 | 97 |