Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000211.4 | NP_000202.3 | 3 - 4 | 377 | 90 |
NM_001127491.2 | NP_001120963.2 | 3 - 4 | 367 | 90 | |
NM_001303238.1 | NP_001290167.1 | 3 - 4 | 332 | 90 | |
XM_006724001.1 | XP_006724064.1 | 3 - 4 | 394 | 90 | |
XM_017028341.1 | XP_016883830.1 | 3 - 4 | 389 | 90 | |
GenBank mRNA | AB208909.1 | - | 3 - 4 | 287 | 90 |
AK095992.1 | - | 3 - 4 | 332 | 90 | |
AK222505.1 | - | 3 - 4 | 234 | 90 | |
AK303488.1 | - | 3 - 4 | 245 | 90 | |
AU279404.1 | - | 4 - 5 | 442 | 90 | |
BC005861.2 | - | 2 - 3 | 188 | 90 | |
BC021077.1 | - | 2 - 3 | 111 | 90 | |
L78790.1 | - | 3 - 4 | 196 | 90 | |
M15395.1 | - | 3 - 4 | 216 | 90 | |
Y00057.1 | - | 2 - 3 | 120 | 90 |