Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005828.4 | NP_005819.3 | 4 - 5 | 764 | 72 |
NR_073585.1 | 4 - 5 | 764 | 72 | ||
GenBank mRNA | BC001264.1 | - | 4 - 5 | 717 | 72 |
BQ221019.1 | - | 4 - 5 | 724 | 72 | |
U94747.1 | - | 4 - 5 | 593 | 72 |