Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317748.1 | NP_001304677.1 | 7 - 8 | 1501 | 78 |
NM_002380.4 | NP_002371.3 | 8 - 9 | 1624 | 78 | |
NM_030583.3 | NP_085072.2 | 8 - 9 | 1624 | 78 | |
XM_005250920.1 | XP_005250977.1 | 8 - 9 | 1589 | 78 | |
XM_017013417.1 | XP_016868906.1 | 7 - 8 | 1466 | 78 | |
XM_017013418.1 | XP_016868907.1 | 8 - 9 | 1589 | 78 | |
GenBank mRNA | AK027775.1 | - | 2 - 3 | 255 | 78 |
AK289721.1 | - | 8 - 9 | 1805 | 78 | |
AL137638.1 | - | 6 - 7 | 686 | 78 | |
AL833931.1 | - | 8 - 9 | 1512 | 78 | |
AY358895.1 | - | 7 - 8 | 1411 | 78 | |
BC010444.1 | - | 8 - 9 | 1553 | 78 | |
BC016394.1 | - | 5 - 6 | 609 | 78 | |
BX649093.1 | - | 5 - 6 | 678 | 78 | |
U69263.2 | - | 7 - 8 | 1456 | 78 |