Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040624.1 | NP_001035714.1 | 7 - 8 | 880 | 109 |
NM_001040625.1 | NP_001035715.1 | 6 - 7 | 793 | 109 | |
NM_001040626.1 | NP_001035716.1 | 6 - 7 | 793 | 109 | |
NM_001040627.1 | NP_001035717.1 | 5 - 6 | 706 | 109 | |
NM_001040628.1 | NP_001035718.1 | 7 - 8 | 849 | 109 | |
NM_001040629.1 | NP_001035719.1 | 6 - 7 | 762 | 109 | |
NM_001040630.1 | NP_001035720.1 | 5 - 6 | 946 | 109 | |
NM_032041.2 | NP_114430.2 | 3 - 4 | 855 | 109 | |
XM_011517332.2 | XP_011515634.1 | 3 - 4 | 644 | 109 | |
XM_011517333.2 | XP_011515635.1 | 7 - 8 | 791 | 109 | |
XM_011517334.2 | XP_011515636.1 | 4 - 5 | 675 | 109 | |
XM_011517335.2 | XP_011515637.1 | 5 - 6 | 767 | 109 | |
XM_017013901.1 | XP_016869390.1 | 6 - 7 | 983 | 109 | |
XM_017013902.1 | XP_016869391.1 | 4 - 5 | 654 | 109 | |
GenBank mRNA | AF251061.1 | - | 5 - 6 | 634 | 109 |
AK055073.1 | - | 3 - 4 | 613 | 109 | |
AK055097.1 | - | 1 - 2 | 1583 | 109 | |
AK096515.1 | - | 7 - 8 | 805 | 109 | |
AK314523.1 | - | 3 - 4 | 614 | 109 | |
AL136612.1 | - | 3 - 4 | 606 | 109 | |
BC036098.1 | - | 5 - 6 | 109 | ||
BC063428.1 | - | 6 - 7 | 779 | 109 |