Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134387.1 | NP_001127859.1 | 4 - 5 | 644 | 154 |
NM_001134388.1 | NP_001127860.1 | 5 - 6 | 553 | 154 | |
NM_001134389.1 | NP_001127861.1 | 4 - 5 | 422 | 154 | |
NM_018106.3 | NP_060576.1 | 4 - 5 | 487 | 154 | |
XM_005249793.2 | XP_005249850.1 | 5 - 6 | 597 | 154 | |
XM_005249794.3 | XP_005249851.1 | 4 - 5 | 587 | 154 | |
XM_005249795.2 | XP_005249852.1 | 4 - 5 | 420 | 154 | |
XM_005249796.3 | XP_005249853.1 | 4 - 5 | 534 | 154 | |
GenBank mRNA | AF201931.1 | - | 3 - 4 | 416 | 154 |
AK001341.1 | - | 4 - 5 | 403 | 154 | |
AK223532.1 | - | 4 - 5 | 487 | 154 | |
AK308971.1 | - | 4 - 5 | 402 | 154 | |
AL136674.1 | - | 4 - 5 | 395 | 154 | |
AY359090.1 | - | 4 - 5 | 575 | 154 | |
BC001239.1 | - | 5 - 6 | 506 | 154 | |
BQ925460.1 | - | 5 - 6 | 513 | 154 | |
CN365375.1 | - | 4 - 5 | 604 | 155 | |
CR533511.1 | - | 2 - 3 | 195 | 154 |