Assay Design



Gene Transcripts

Gene Symbol
ZDHHC4
Entrez Gene ID
55146
Gene Name
zinc finger DHHC-type containing 4
Gene Aliases
ZNF374
Chromosome Location
Chr.7: 6577413 - 6588996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134387.1 NP_001127859.1 1 - 2 293 73
NM_001134388.1 NP_001127860.1 2 - 3 202 73
XM_005249794.3 XP_005249851.1 1 - 2 210 73
XM_017012391.1 XP_016867880.1 1 - 2 211 73
GenBank mRNA AY359090.1 - 1 - 2 224 73
BC001239.1 - 2 - 3 155 73
BQ925460.1 - 2 - 3 162 73
CN365375.1 - 1 - 2 252 73

View Full Product Details