Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018164.2 | NP_060634.2 | 2 - 3 | 764 | 102 |
XM_005253422.3 | XP_005253479.1 | 2 - 3 | 723 | 102 | |
XM_011520750.2 | XP_011519052.1 | 2 - 3 | 297 | 102 | |
XM_011520751.2 | XP_011519053.1 | 2 - 3 | 723 | 102 | |
XM_011520752.2 | XP_011519054.1 | 2 - 3 | 723 | 102 | |
XM_017019632.1 | XP_016875121.1 | 2 - 3 | 297 | 102 | |
XM_017019635.1 | XP_016875124.1 | 2 - 3 | 297 | 102 | |
XM_017019636.1 | XP_016875125.1 | 2 - 3 | 723 | 102 | |
GenBank mRNA | AK001492.1 | - | 2 - 3 | 393 | 102 |
AK225336.1 | - | 2 - 3 | 415 | 102 | |
BC003081.2 | - | 2 - 3 | 453 | 102 | |
BC008368.1 | - | 2 - 3 | 411 | 102 |