Assay Design



Gene Transcripts

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
Chromosome Location
Chr.22: 29327680 - 29388583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127.3 NP_001118.3 1 - 2 159 93
NM_001166019.1 NP_001159491.1 1 - 2 159 93
NM_145730.2 NP_663782.2 1 - 2 159 93
GenBank mRNA BC046242.1 - 1 - 2 159 93
BM909530.1 - 1 - 2 156 93

View Full Product Details